Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016870.3 | NP_058566.3 | 15 | 2036 | ||
NM_212441.2 | NP_997606.2 | 14 | 1931 | |||
NM_212442.2 | NP_997607.2 | 14 | 1834 | |||
GenBank mRNA | AB022340.2 | - | 14 | 1822 | ||
AF068246.1 | - | 14 | 1821 | |||
AK077247.1 | - | 14 | 1897 | |||
AK143946.1 | - | 1834 | ||||
AK165516.1 | - | 14 | 1808 | |||
AY064696.1 | - | 14 | 1907 | |||
BC015248.1 | - | 15 | 1996 | 4239288 |