Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016875.2 | NP_058571.2 | 9 | 1366 | ||
GenBank mRNA | AF073954.1 | - | 9 | 1293 | ||
AF073955.1 | - | 9 | 1160 | |||
AK131732.1 | - | 3 | 485 | |||
AK131786.1 | - | 9 | 1345 | |||
AK161439.1 | - | 5 | 758 | |||
BC100454.1 | - | 6 | 746 |