Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016876.3 | NP_058572.2 | 5, 6 | 328 | ||
GenBank mRNA | AF108214.1 | - | 5, 6 | 342 | ||
AK038249.1 | - | 5, 6 | 338 | |||
AK168362.1 | - | 5, 6 | 331 | |||
BC008511.1 | - | 5, 6 | 328 | 3592537 | ||
U70733.1 | - | 5, 6 | 318 |