Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 9, 10 | 1127 | ||
NM_016878.4 | NP_058574.3 | 9, 10 | 951 | |||
GenBank mRNA | AF005051.1 | - | 8, 9 | 1061 | ||
AK038646.1 | - | 1127 | ||||
AK052001.1 | - | 929 | ||||
AK160084.1 | - | 9, 10 | 888 | |||
AK166276.1 | - | 9, 10 | 901 | |||
AK215853.1 | - | 2, 3 | 188 | |||
BC092232.1 | - | 9, 10 | 879 | 30551654 |