Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012128.3 | NP_036260.2 | 1, 2 | 211 | ||
NR_024602.1 | 1, 2 | 211 | ||||
XM_011541015.2 | XP_011539317.1 | 1, 2 | 30 | |||
GenBank mRNA | AF127035.1 | - | 1, 2 | 195 | ||
AK000072.1 | - | 1, 2 | 209 | |||
AK293766.1 | - | 1, 2 | 44 | |||
AK307785.1 | - | 1, 2 | 155 | |||
AK307898.1 | - | 1, 2 | 72 | |||
AK314430.1 | - | 1, 2 | 207 | |||
AY358470.1 | - | 1, 2 | 191 | |||
BC113687.1 | - | 1, 2 | 206 | 8322739 | ||
BC113689.1 | - | 1, 2 | 205 | 8322741 | ||
DA761219.1 | - | 1, 2 | 273 |