Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016892.3 | NP_058588.1 | 6, 7 | 647 | ||
GenBank mRNA | AF121906.1 | - | 6, 7 | 560 | ||
AF173379.1 | - | 6, 7 | 620 | |||
AK010264.1 | - | 3, 4 | 256 | |||
AK050072.1 | - | 6, 7 | 596 | |||
BC026938.1 | - | 6, 7 | 614 | 4024140 |