Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016904.1 | NP_058600.1 | 3 | 438 | ||
GenBank mRNA | AB025409.1 | - | 3 | 438 | ||
AK004101.1 | - | 3 | 424 | |||
AK010414.2 | - | 3 | 425 | |||
AK011314.1 | - | 3 | 427 | |||
AK011782.1 | - | 3 | 404 | |||
AK019429.1 | - | 1 | 92 | |||
BC076579.1 | - | 3 | 430 | 6846517 |
Gene Symbol: Cks1brt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037922.3 | NP_001033011.1 | 2 | 709 | ||
GenBank mRNA | AK144293.1 | - | 2 | 710 | ||
BC147100.1 | - | 1 | 606 | 9056144 | ||
BC147101.1 | - | 1 | 606 | 9056154 |