Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016906.4 | NP_058602.1 | 12 | 1913 | ||
GenBank mRNA | AK007696.1 | - | 1 | 275 | ||
AK077849.1 | - | 1 | 422 | |||
AK088473.1 | - | 12 | 1914 | |||
AK153055.1 | - | 12 | 1903 | |||
AK159349.1 | - | 12 | 1899 | |||
AK167484.1 | - | 12 | 1842 | |||
BC003707.1 | - | 12 | 1839 | 3592772 | ||
BC006020.1 | - | 8 | 1498 |