Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsnax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016909.2 | NP_058605.1 | 3 | 414 | ||
GenBank mRNA | AF187040.1 | - | 3 | 414 | ||
AK014437.1 | - | 3 | 362 | |||
AK030407.1 | - | 3 | 344 | |||
AK146339.1 | - | 3 | 347 | |||
AK159223.1 | - | 3 | 416 | |||
AK164453.1 | - | 2 | 189 | |||
AK189358.1 | - | 3 | 219 | |||
BC004611.1 | - | 3 | 316 | 3585400 |