Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gal3st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177691.1 | NP_001171162.1 | 3 | 1359 | ||
NM_001177703.1 | NP_001171174.1 | 3 | 1490 | |||
NM_016922.3 | NP_058618.2 | 4 | 1708 | |||
GenBank mRNA | AB032939.1 | - | 2 | 1285 | ||
AK002510.1 | - | 3 | 1355 | |||
AK007645.1 | - | 3 | 1490 | |||
BB804195.1 | - | 1 | 104 | |||
BC026806.1 | - | 4 | 1631 | 4215616 |