Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016925.3 | NP_058621.2 | 31 | 3050 | ||
GenBank mRNA | AF178934.1 | - | 31 | 3046 | ||
AF208116.1 | - | 31 | 3052 | |||
AF208120.1 | - | 10 | 1133 | |||
BC150705.1 | - | 31 | 3048 | 9087616 |