Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016963.2 | NP_058659.1 | 10 | 1241 | ||
GenBank mRNA | AF177172.1 | - | 10 | 1229 | ||
AF237630.1 | - | 10 | 1155 | |||
AK017725.1 | - | 10 | 1237 | |||
AK050372.1 | - | 10 | 1243 | |||
AK084463.1 | - | 7 | 763 | |||
AK147017.1 | - | 10 | 1242 | |||
AK167723.1 | - | 10 | 1239 | |||
AK169218.1 | - | 10 | 1242 | |||
BC082595.1 | - | 10 | 1233 | 30645500 |