Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016971.2 | NP_058667.1 | 5 | 508 | ||
GenBank mRNA | AJ249491.1 | - | 5 | 534 | ||
BC104348.1 | - | 5 | 508 | 40049545 | ||
BC104349.1 | - | 5 | 508 | 40049546 | ||
BC116235.2 | - | 5 | 504 | 40091712 |