Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mc4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016977.4 | NP_058673.2 | 1 | 570 | ||
GenBank mRNA | AK136793.1 | - | 1 | 556 | ||
BC116957.1 | - | 1 | 188 | 40126276 | ||
BC116959.1 | - | 1 | 188 | 40126278 |
Set Membership: |
G-protein coupled receptor |