Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017367.3 | NP_059063.2 | 2 | 718 | ||
GenBank mRNA | AF005886.2 | - | 2 | 370 | ||
AK050465.1 | - | 2 | 719 | |||
AK152614.1 | - | 2 | 675 | |||
AK212895.1 | - | 2 | 176 | |||
BC003290.1 | - | 2 | 251 | 3583418 | ||
BC029328.1 | - | 2 | 251 |