Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168568.1 | NP_001162040.1 | 4 | 601 | ||
NM_001168569.1 | NP_001162041.1 | 4 | 658 | |||
NM_001168571.1 | NP_001162043.1 | 4 | 733 | |||
NM_018737.5 | NP_061207.1 | 4 | 733 | |||
GenBank mRNA | AK032018.1 | - | 4 | 460 | ||
AK052854.1 | - | 4 | 649 | |||
AK084595.1 | - | 4 | 530 | |||
AK085381.1 | - | 4 | 508 | |||
AK150966.1 | - | 4 | 600 | |||
AK151597.1 | - | 4 | 600 | |||
AK161564.1 | - | 4 | 571 | |||
AK164206.1 | - | 4 | 560 | |||
AK178727.1 | - | 2 | 66 | |||
BC003257.1 | - | 4 | 441 | 3500778 | ||
U49385.1 | - | 4 | 623 |