Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253700.1 | NP_001240629.1 | 2 | 160 | ||
NM_018740.2 | NP_061210.2 | 3 | 421 | |||
GenBank mRNA | AA646690.1 | - | 2 | 76 | ||
AF004107.1 | - | 2 | 340 | |||
AK011130.1 | - | 1 | 233 | |||
AK013662.1 | - | 1 | 22 | |||
AK152582.1 | - | 3 | 425 | |||
BC003752.1 | - | 2 | 272 | 3489878 | ||
BY292587.1 | - | 1 | 223 |