Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016933.3 | NP_058629.2 | 2 | 817 | ||
GenBank mRNA | AI627045.1 | - | 1 | 453 | ||
AK088334.1 | - | 2 | 814 | |||
AK165203.1 | - | 2 | 815 | |||
BC013273.1 | - | 2 | 816 | 4208992 | ||
BC100560.1 | - | 2 | 798 | 832557 | ||
D10105.1 | - | 2 | 793 | |||
U03856.1 | - | 1 | 810 |