Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016978.2 | NP_058674.1 | 2, 3 | 347 | ||
GenBank mRNA | AK128914.1 | - | 2, 3 | 338 | ||
AK145877.1 | - | 2, 3 | 348 | |||
AK146413.1 | - | 2, 3 | 292 | |||
AK168851.1 | - | 2, 3 | 296 | |||
AK185243.1 | - | 1, 2 | 131 | |||
AK185430.1 | - | 1, 2 | 131 | |||
BC008119.1 | - | 2, 3 | 280 | 3498240 | ||
X64837.1 | - | 1, 2 | 321 |