Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016979.1 | NP_058675.1 | 2 | 470 | ||
GenBank mRNA | AK034681.1 | - | 2 | 789 | ||
AK036432.1 | - | 2 | 435 | |||
AK037141.1 | - | 2 | 464 | |||
AK039088.1 | - | 2 | 485 | |||
AK081548.1 | - | 2 | 450 | |||
AK139510.1 | - | 2 | 478 | |||
AK149937.1 | - | 2 | 448 | |||
AK150588.1 | - | 2 | 432 | |||
AK152166.1 | - | 2 | 267 | |||
AK154447.1 | - | 2 | 450 | |||
AK169322.1 | - | 2 | 421 | |||
BC006875.1 | - | 2 | 470 | 3596013 |