Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017367.3 | NP_059063.2 | 4, 5 | 943 | ||
GenBank mRNA | AF005886.2 | - | 4, 5 | 595 | ||
AK050465.1 | - | 4, 5 | 944 | |||
AK079476.1 | - | 4, 5 | 884 | |||
AK152614.1 | - | 4, 5 | 902 | |||
AK208767.1 | - | 2, 3 | 216 | |||
BC003290.1 | - | 4, 5 | 476 | 3583418 | ||
BC029328.1 | - | 4, 5 | 476 |