Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256796.1 | NP_001243725.1 | 7, 8 | 968 | ||
NM_001261397.1 | NP_001248326.1 | 6, 7 | 839 | |||
NM_001261398.1 | NP_001248327.1 | 6, 7 | 920 | |||
NM_002560.2 | NP_002551.2 | 6, 7 | 920 | |||
NR_046372.1 | 7, 8 | 958 | ||||
NR_046373.1 | 5, 6 | 772 | ||||
XM_011538416.2 | XP_011536718.1 | 2, 3 | 283 | |||
GenBank mRNA | AF000234.1 | - | 6, 7 | 516 | ||
AF012903.1 | - | 5, 6 | 402 | |||
AK225604.1 | - | 6, 7 | 616 | |||
AK293599.1 | - | 3, 4 | 410 | |||
AK293660.1 | - | 5, 6 | 555 | |||
AK297704.1 | - | 5, 6 | 504 | |||
AK301080.1 | - | 6, 7 | 604 | |||
AK308549.1 | - | 7, 8 | 690 | |||
AK308868.1 | - | 6, 7 | 1611 | |||
BC017458.1 | - | 6, 7 | 649 | |||
BC033826.1 | - | 6, 7 | 640 | 5216449 | ||
BT019738.1 | - | 6, 7 | 616 | |||
BT019739.1 | - | 6, 7 | 535 | |||
BX402887.2 | - | 7, 8 | 738 | |||
DQ851491.1 | - | 2, 3 | 134 | |||
U83993.1 | - | 6, 7 | 925 | |||
Y07684.1 | - | 6, 7 | 643 |