Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002562.5 | NP_002553.3 | 2 | 370 | ||
NR_033948.1 | 2 | 370 | ||||
NR_033949.1 | 2 | 370 | ||||
NR_033950.1 | 2 | 370 | ||||
NR_033951.1 | 2 | 370 | ||||
NR_033952.1 | 2 | 370 | ||||
NR_033954.1 | 2 | 370 | ||||
NR_033955.1 | 2 | 370 | ||||
NR_033956.1 | 2 | 370 | ||||
XM_017019364.1 | XP_016874853.1 | 2 | 370 | |||
XM_017019366.1 | XP_016874855.1 | 2 | 311 | |||
GenBank mRNA | AB209709.1 | - | 2 | 291 | ||
AK290405.1 | - | 2 | 318 | |||
AK294126.1 | - | 2 | 321 | |||
AK296432.1 | - | 2 | 318 | |||
AY847298.1 | - | 2 | 227 | |||
AY847299.1 | - | 2 | 227 | |||
AY847300.1 | - | 2 | 227 | |||
AY847301.1 | - | 2 | 227 | |||
AY847302.1 | - | 2 | 227 | |||
AY847303.1 | - | 2 | 227 | |||
AY847304.1 | - | 2 | 227 | |||
BC007679.2 | - | 2 | 268 | 3628076 | ||
BC011913.2 | - | 2 | 296 | 4298811 | ||
BC121157.1 | - | 2 | 296 | |||
BC121158.1 | - | 2 | 296 | |||
BI598073.1 | - | 2 | 360 | |||
DA274887.1 | - | 2 | 256 | |||
DQ399293.1 | - | 2 | 227 | |||
GQ180122.1 | - | 2 | 236 | |||
Y09561.1 | - | 2 | 253 |