Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017401.2 | NP_059097.2 | 5, 6 | 971 | ||
GenBank mRNA | AF176098.1 | - | 5, 6 | 744 | ||
AJ251804.2 | - | 5, 6 | 715 | |||
BC056213.1 | - | 5, 6 | 763 | 30024844 | ||
BC056996.1 | - | 2, 3 | 2618 |