Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017404.4 | NP_059100.3 | 7 | 807 | ||
GenBank mRNA | AF239728.1 | - | 7 | 724 | ||
AK007681.1 | - | 7 | 743 | |||
AK031942.1 | - | 7 | 728 | |||
AK209039.1 | - | 2 | 72 | |||
BC012274.1 | - | 7 | 686 | 3981658 | ||
BC016561.1 | - | 7 | 743 | 2645316 | ||
BC036276.1 | - | 7 | 743 |