Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111324.2 | NP_001104794.1 | 3 | 706 | ||
NM_017464.5 | NP_059492.3 | 2 | 487 | |||
GenBank mRNA | AF009366.1 | - | 3 | 642 | ||
AK030985.1 | - | 3 | 640 | |||
AK033729.1 | - | 2 | 488 | |||
AK046357.1 | - | 2 | 450 | |||
AK054179.1 | - | 2 | 483 | |||
AK083374.1 | - | 3 | 634 | |||
BC053713.1 | - | 3 | 639 | 30077589 |