Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018739.2 | NP_061209.1 | 4 | 485 | ||
GenBank mRNA | AK146136.1 | - | 4 | 466 | ||
AK147912.1 | - | 488 | ||||
AK168391.1 | - | 4 | 478 | |||
BC048480.1 | - | 4 | 343 | |||
BC115549.1 | - | 4 | 445 | 40101700 | ||
BC115550.1 | - | 4 | 445 | 40101701 | ||
D78255.1 | - | 4 | 464 |