Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253700.1 | NP_001240629.1 | 3 | 242 | ||
NM_018740.2 | NP_061210.2 | 4 | 503 | |||
GenBank mRNA | AA646690.1 | - | 3 | 158 | ||
AF004107.1 | - | 3 | 422 | |||
AK011130.1 | - | 2 | 315 | |||
AK013662.1 | - | 2 | 104 | |||
AK152582.1 | - | 4 | 507 | |||
AK199980.1 | - | 1 | 49 | |||
BC003752.1 | - | 3 | 354 | 3489878 | ||
BY292587.1 | - | 2 | 305 |