Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102458.1 | NP_001095928.1 | 6, 7 | 1208 | ||
NM_001301688.1 | NP_001288617.1 | 5, 6 | 1048 | |||
NM_018745.5 | NP_061215.1 | 5, 6 | 1048 | |||
GenBank mRNA | AF032128.1 | - | 3, 4 | 474 | ||
AK049680.1 | - | 5, 6 | 1049 | |||
AK088112.1 | - | 5, 6 | 1046 | |||
AK088671.1 | - | 6, 7 | 1205 | |||
AK133914.1 | - | 5, 6 | 1044 | |||
BC019412.1 | - | 5, 6 | 1036 | 5151406 | ||
BC021152.1 | - | 5, 6 | 1001 | |||
BC043722.1 | - | 3, 4 | 554 | 5321733 | ||
CF738062.1 | - | 1, 2 | 39 |