Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016978.2 | NP_058674.1 | 4 | 632 | ||
GenBank mRNA | AK128914.1 | - | 4 | 623 | ||
AK145877.1 | - | 4 | 633 | |||
AK146413.1 | - | 4 | 577 | |||
AK168851.1 | - | 4 | 581 | |||
AK187824.1 | - | 2 | 275 | |||
BC008119.1 | - | 4 | 565 | 3498240 | ||
X64837.1 | - | 3 | 606 |