Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017372.3 | NP_059068.1 | 2 | 307 | ||
GenBank mRNA | AK148516.1 | - | 2 | 308 | ||
AK150998.1 | - | 2 | 310 | |||
AK153244.1 | - | 2 | 310 | |||
AK153475.1 | - | 2 | 308 | |||
AK159276.1 | - | 2 | 357 | |||
AK159640.1 | - | 2 | 304 | |||
BC002069.1 | - | 2 | 299 | 3593360 | ||
BC054463.1 | - | 2 | 299 | 6514977 |