Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017376.3 | NP_059072.1 | 4 | 2958 | ||
NM_153484.3 | NP_705617.2 | 4 | 2865 | |||
GenBank mRNA | AK009518.1 | - | 1 | 456 | ||
AK042146.1 | - | 4 | 3119 | |||
AK085437.1 | - | 1 | 106 | |||
AK141606.1 | - | 4 | 2786 | |||
AK147965.1 | - | 4 | 2893 | |||
AK154472.1 | - | 4 | 2903 | |||
AK178639.1 | - | 1 | 173 | |||
AK193624.1 | - | 1 | 172 | |||
AK201843.1 | - | 1 | 172 | |||
AK208083.1 | - | 1 | 196 | |||
AK220090.1 | - | 1 | 172 | |||
AY540631.1 | - | 4 | 2943 | |||
AY540632.1 | - | 4 | 2757 | |||
BC017689.1 | - | 4 | 2721 | 4242534 | ||
BC023410.1 | - | 1 | 1389 | |||
BC036982.1 | - | 4 | 2923 | 4976241 |