Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017379.1 | NP_059075.1 | 3 | 363 | ||
GenBank mRNA | AJ245923.1 | - | 3 | 363 | ||
AK032157.1 | - | 336 | ||||
BC017631.1 | - | 3 | 342 | 4507364 | ||
GU591980.1 | - | 315 |