Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017399.4 | NP_059095.1 | 3 | 306 | ||
GenBank mRNA | AK131868.1 | - | 3 | 316 | ||
BC009812.1 | - | 3 | 306 | 4159971 | ||
Y14660.1 | - | 3 | 306 |