Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102458.1 | NP_001095928.1 | 13 | 2448 | ||
NM_001301688.1 | NP_001288617.1 | 12 | 2040 | |||
NM_018745.5 | NP_061215.1 | 12 | 2288 | |||
GenBank mRNA | AK049680.1 | - | 12 | 2289 | ||
AK088112.1 | - | 12 | 2285 | |||
AK088671.1 | - | 13 | 2444 | |||
AK133914.1 | - | 12 | 2284 | |||
BC019412.1 | - | 12 | 2275 | 5151406 | ||
BC021152.1 | - | 12 | 2240 | |||
BC043722.1 | - | 10 | 1794 | 5321733 | ||
BC046814.1 | - | 12 | 2340 | |||
CF749918.1 | - | 3 | 493 |