Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018761.3 | NP_061231.3 | 2, 3 | 433 | ||
GenBank mRNA | AF006071.1 | - | 2, 3 | 420 | ||
AK034393.1 | - | 2, 3 | 390 | |||
AK082755.1 | - | 2, 3 | 376 | |||
BC050070.1 | - | 2, 3 | 410 | 30012833 | ||
CF896873.1 | - | 2, 3 | 360 | |||
CJ094037.1 | - | 2, 3 | 434 |