Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018771.3 | NP_061241.1 | 2 | 324 | ||
GenBank mRNA | AF061263.1 | - | 1 | 258 | ||
AF089818.1 | - | 1 | 281 | |||
AF104358.1 | - | 2 | 302 | |||
AK002492.1 | - | 2 | 305 | |||
AK086506.1 | - | 2 | 325 | |||
AK183407.1 | - | 1 | 17 | |||
BC003490.1 | - | 2 | 324 | 2811749 |