Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018777.4 | NP_061247.1 | 3 | 326 | ||
GenBank mRNA | AF087824.1 | - | 1 | 203 | ||
AF125305.1 | - | 1 | 203 | |||
AK010560.1 | - | 3 | 327 | |||
AK010682.1 | - | 3 | 321 | |||
BC005718.1 | - | 3 | 303 | 3594823 | ||
BC050138.1 | - | 3 | 309 | 30028814 | ||
DQ864649.1 | - | 1 | 203 |