Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018789.2 | NP_061259.1 | 1 | 718 | ||
GenBank mRNA | AB032770.1 | - | 1 | 498 | ||
AB221664.1 | - | 1 | 425 | |||
AF114260.1 | - | 1 | 718 | |||
AK157830.1 | - | 1 | 680 | |||
BC099460.1 | - | 1 | 657 | |||
BC139275.1 | - | 1 | 559 | 8862297 | ||
BC139304.1 | - | 1 | 559 | 8862326 |