Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205312.1 | NP_001192241.1 | 3 | 367 | ||
NM_018793.2 | NP_061263.2 | 3 | 367 | |||
GenBank mRNA | AF173032.1 | - | 3 | 302 | ||
AK154438.1 | - | 3 | 325 | |||
AK155923.1 | - | 3 | 280 | |||
AK159303.1 | - | 3 | 336 | |||
AK165483.1 | - | 3 | 336 | |||
AK167726.1 | - | 3 | 346 | |||
AK170107.1 | - | 3 | 306 | |||
AK170940.1 | - | 3 | 306 | |||
AK171609.1 | - | 3 | 280 | |||
BC053021.1 | - | 2 | 272 | |||
BC094240.1 | - | 2 | 263 | 30610201 |