Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018824.2 | NP_061294.2 | 4 | 509 | ||
GenBank mRNA | AB038145.1 | - | 3 | 245 | ||
AK087175.1 | - | 4 | 517 | |||
AK169991.1 | - | 4 | 511 | |||
AY004874.1 | - | 4 | 463 | |||
BC050823.1 | - | 3 | 273 | 6332451 |