Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207678.1 | NP_997561.1 | 10, 11 | 1249 | ||
GenBank mRNA | AB041605.1 | - | 10, 11 | 1226 | ||
AK048244.1 | - | 10, 11 | 3029 | |||
AY337018.1 | - | 10, 11 | 1232 | |||
BC003773.1 | - | 3, 4 | 372 | |||
BC023747.1 | - | 10, 11 | 3017 | |||
BC083055.1 | - | 3, 4 | 236 | |||
BC132295.1 | - | 10, 11 | 1241 | 40130572 | ||
U37351.1 | - | 3, 4 | 255 |