Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018748.3 | NP_061218.2 | 2 | 289 | ||
GenBank mRNA | AF051357.2 | - | 2 | 288 | ||
AK161568.1 | - | 2 | 341 | |||
AK172292.1 | - | 2 | 337 | |||
AK219816.1 | - | 1 | 36 | |||
BC003268.1 | - | 2 | 289 | |||
BC023102.1 | - | 2 | 224 | |||
BC037641.1 | - | 2 | 334 | |||
BC057561.1 | - | 2 | 156 | |||
BC096467.1 | - | 2 | 289 |