Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018750.3 | NP_061220.2 | 4 | 753 | ||
GenBank mRNA | AF053959.1 | - | 4 | 735 | ||
AK005059.2 | - | 4 | 749 | |||
AK029321.1 | - | 4 | 755 | |||
AK088751.1 | - | 3 | 388 | |||
AK131147.1 | - | 3 | 346 | |||
AK155534.1 | - | 4 | 749 | |||
AK155869.1 | - | 3 | 425 | |||
AY261333.1 | - | 3 | 364 | |||
BC089605.1 | - | 4 | 755 | 30121804 |