Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018753.6 | NP_061223.2 | 2 | 398 | ||
GenBank mRNA | AK002632.1 | - | 2 | 417 | ||
AK004872.1 | - | 2 | 399 | |||
AK011389.1 | - | 2 | 398 | |||
AK083367.1 | - | 2 | 388 | |||
AK144061.1 | - | 2 | 461 | |||
AK150414.1 | - | 2 | 411 | |||
AK151294.1 | - | 2 | 411 | |||
AK158932.1 | - | 2 | 301 | |||
BC049070.1 | - | 232 | ||||
BC132121.1 | - | 2 | 252 | 40130398 | ||
BC138512.1 | - | 2 | 252 | 8861532 |