Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323971.1 | NP_001310900.1 | 2 | 528 | ||
NM_001323973.1 | NP_001310902.1 | 2 | 528 | |||
NM_001323974.1 | NP_001310903.1 | 2 | 528 | |||
NM_001323975.1 | NP_001310904.1 | 2 | 528 | |||
NM_004594.2 | NP_004585.1 | 2 | 448 | |||
NR_136664.1 | 2 | 528 | ||||
NR_136665.1 | 2 | 528 | ||||
NR_136666.1 | 2 | 528 | ||||
NR_136667.1 | 2 | 528 | ||||
XM_017023594.1 | XP_016879083.1 | 2 | 1851 | |||
GenBank mRNA | AB209203.1 | - | 2 | 972 | ||
AF111173.1 | - | 2 | 454 | |||
BC142671.1 | - | 2 | 411 | 40148180 | ||
BC144131.1 | - | 2 | 528 | |||
BC150207.1 | - | 2 | 423 | |||
DB475730.1 | - | 2 | 452 |