Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crlf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277106.1 | NP_001264035.1 | 2 | 222 | ||
NM_018776.2 | NP_061246.1 | 2 | 222 | |||
GenBank mRNA | AF046060.1 | - | 2 | 197 | ||
AF120152.1 | - | 2 | 131 | |||
AK037880.1 | - | 2 | 170 | |||
AK078569.1 | - | 2 | 167 | |||
AK137528.1 | - | 2 | 174 | |||
AK146798.1 | - | 2 | 170 | |||
AK165640.1 | - | 2 | 172 | |||
BC004780.1 | - | 2 | 169 | 3257685 |