Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018782.2 | NP_061252.2 | 7 | 763 | ||
GenBank mRNA | AB015595.1 | - | 7 | 767 | ||
AF146525.1 | - | 5 | 440 | |||
AF209905.1 | - | 3 | 259 | |||
AK137210.1 | - | 7 | 762 | |||
BC138537.1 | - | 4 | 390 | 8861557 | ||
BC138540.1 | - | 4 | 390 | 8861560 |
Set Membership: |
G-protein coupled receptor |