Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018786.2 | NP_061256.1 | 8 | 530 | ||
GenBank mRNA | AF135440.1 | - | 8 | 530 | ||
AK140075.1 | - | 8 | 699 | |||
AK164947.1 | - | 9 | 743 | |||
AK217476.1 | - | 2 | 145 | |||
BC051961.1 | - | 7 | 509 | |||
BC058389.1 | - | 6 | 1434 | |||
BC082994.1 | - | 8 | 544 | 6333535 |