Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018814.3 | NP_061284.2 | 12 | 3438 | ||
GenBank mRNA | AF096286.1 | - | 6 | 1268 | ||
AK129221.1 | - | 7 | 1318 | |||
AK135724.1 | - | 6 | 894 | |||
BC057333.1 | - | 5 | 1181 | |||
BC066849.1 | - | 7 | 1111 | |||
BC098370.1 | - | 7 | 1028 |